Learn More
Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed, Thermo Scientific Chemicals
Form: approx. 40.1g/150 x 300mm
Brand: Thermo Scientific Alfa Aesar 045092.4K
Code : NEW
Additional Details : Weight : 0.01000kg
Beschreibung
It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Nickel Iron Molybdenum foil | |
Ni:Fe:Mo; 80:15:4.2 wt% | |
MFCD09910537 | |
Insoluble in water. | |
Foil |
0.102mm (0.004 in.) thick, Annealed | |
Ni:Fe:Mo | |
150 x 300 mm | |
≈40.1g/150x300mm |
RUO – Research Use Only