Learn More
Nickel Iron Copper Molybdenum foil, 0.125mm (0.005 in.) thick, annealed (Mumetal™)
Description
Nickel Iron Copper Molybdenum foil is used as a magnetic shielding alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nickel Iron Copper Molybdenum foil |
| Name Note | 0.125mm (0.005 in.) thick, Annealed (Mumetal™) |
| Avg. Mol. Wt. or Mol. Wt. Range | Ni:Fe:Cu:Mo; 77:14:5:4 wt% |
| Quantity | 75 x 75 mm |
| Form | Foil |
| MDL Number | MFCD02684505 |
| Solubility Information | Insoluble in water. |
Safety and Handling
- Nickel Iron Copper Molybdenum foil, 0.125mm (0.005in) thick, annealed
- Danger
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: Nickel
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.