missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) tetrafluoroborate hexahydrate

Product Code. 11458650
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458650 500 g 500g
11448650 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458650 Supplier Thermo Scientific Alfa Aesar Supplier No. 053108.36
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol

It is mainly used as an electroplating intermediate. It is also used in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15684-36-3
Molecular Formula B2F8H12NiO6
Molecular Weight (g/mol) 340.39
MDL Number MFCD00150259
InChI Key BFFWGFMAXHCLDV-UHFFFAOYSA-N
Synonym nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate
PubChem CID 177615
SMILES O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Physical Form Lump
Quantity 500 g
UN Number UN3077
Solubility Information Soluble in water.
IUPAC Name nickel(2+);ditetrafluoroborate;hexahydrate
Formula Weight 340.40 (232.30 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.5 g/mL
Chemical Name or Material Nickel (II) tetrafluoroborate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.