missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) tetrafluoroborate hexahydrate, 99%

Product Code. 10405862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10405862 25 g Glass bottle 25g
10621073 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10405862 Supplier Thermo Scientific Acros Supplier No. 358730250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15684-36-3 , 14708-14-6
Molecular Formula B2F8H12NiO6
Molecular Weight (g/mol) 340.39
MDL Number MFCD00150259
InChI Key BFFWGFMAXHCLDV-UHFFFAOYSA-N
Synonym nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate
PubChem CID 177615
SMILES O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue-Green
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystals
Quantity 25 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ni(BF4)2·6H2O
Solubility Information Solubility in water: soluble in water
IUPAC Name nickel(2+);ditetrafluoroborate;hexahydrate
Formula Weight 340.39
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Nickel(II) tetrafluoroborate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) tetrafluoroborate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.