missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hexahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog Number 11353418
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10101-97-0
H12NiO10S
262.839
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
10101-97-0
H12NiO10S
262.839
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

It is the precursor material for Ni electroplating. It is applied in biochemistry and also employed as a calibrant for magnetic susceptibility measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min.
Solubility Information Soluble in water. Slightly soluble in alcohol and methanol
Formula Weight 262.86 (154.77 Anhydrous)
Grade ACS
Density 2.07
Chemical Name or Material Nickel (II) Sulfate Hexahydrate, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'