Learn More
Nickel(II) sulfate hexahydrate, ACS, 98.0% min, Thermo Scientific Chemicals
£30.00 - £260.00
Chemical Identifiers
CAS | 10101-97-0 |
---|---|
Molecular Formula | H12NiO10S |
Molecular Weight (g/mol) | 262.839 |
MDL Number | MFCD00149813 |
InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
IUPAC Name | nickel(2+);sulfate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353418
|
Thermo Scientific Alfa Aesar
036336.22 |
100 g |
£30.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343418
|
Thermo Scientific Alfa Aesar
036336.36 |
500 g |
£70.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363418
|
Thermo Scientific Alfa Aesar
036336.A4 |
2.5 kg |
£260.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is the precursor material for Ni electroplating. It is applied in biochemistry and also employed as a calibrant for magnetic susceptibility measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-97-0 | |
262.839 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
nickel(2+);sulfate;hexahydrate |
H12NiO10S | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
10101-97-0 | |
Powder | |
H12NiO10S | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.86 (154.77 Anhydrous) | |
2.07 |
Blue | |
98% min. | |
MFCD00149813 | |
Soluble in water. Slightly soluble in alcohol and methanol | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.839 | |
CHEBI:53437 | |
ACS | |
Nickel (II) Sulfate Hexahydrate, For analysis ACS |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372
missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 232-104-9
missing translation for 'rtecsNumber' : QR9600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only