missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Sulfate Hexahydrate, ≥99.99% Trace Metals Basis, Honeywell™
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | 53°C |
Color | Green |
pH | 4 to 6 (at 20°C) |
Physical Form | Solid |
Quantity | 250 g |
Linear Formula | NiSO4 · 6H2O |
UN Number | UN3077 |
Solubility Information | In water 378g/L at 20°C |
Formula Weight | 262.85g/mol |
Percent Purity | ≥99.99% (trace metals basis) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'