missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfamate hydrate

Product Code. 11329369
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329369 1 kg 1kg
11339369 50 g 50g
11349369 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329369 Supplier Thermo Scientific Alfa Aesar Supplier No. 089020.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13770-89-3 | H4N2NiO6S2 | 250.85 g/mol

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-89-3
Molecular Formula H4N2NiO6S2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00137261
InChI Key KERTUBUCQCSNJU-UHFFFAOYSA-L
Synonym nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
PubChem CID 83720
SMILES [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystals
Quantity 1 kg
Solubility Information Soluble in water.
IUPAC Name nickel(2+);disulfamate
Formula Weight 250.86 (Anhydrous)
Odor Odorless
Chemical Name or Material Nickel(II) sulfamate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.