missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) perchlorate hydrate, 99.998% (metals basis)

Product Code. 11418420
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418420 10 g 10g
11428420 50 g 50g
11438420 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418420 Supplier Thermo Scientific Alfa Aesar Supplier No. 044316.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13637-71-3 | Cl2NiO8 | 257.59 g/mol

Applied as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13637-71-3
Molecular Formula Cl2NiO8
Molecular Weight (g/mol) 257.59
MDL Number MFCD00149804
InChI Key ZLQBNKOPBDZKDP-UHFFFAOYSA-L
Synonym nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
PubChem CID 26158
SMILES [Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 140°C
Color Green
Physical Form Crystalline
Quantity 10 g
Linear Formula Ni(ClO4)2·xH2O
UN Number UN1481
Solubility Information Very soluble in water. Soluble in many organic solvents.
IUPAC Name nickel(2+) diperchlorate
Formula Weight 257.59 (Anhydrous)
Percent Purity 99.998%
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Nickel (II) perchlorate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.