missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, black, 76% Ni, Thermo Scientific Chemicals

£22.60 - £574.00

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10501091
View Documents Promotion Details
Thermo Scientific Acros
223120050
5 g Glass bottle
£22.60
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10318550
View Documents Promotion Details
Thermo Scientific Acros
223121000
100 g Glass bottle
£32.10
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10471261
View Documents Promotion Details
Thermo Scientific Acros
223125000
500 g Glass bottle
£136.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10300744
View Documents Promotion Details
Thermo Scientific Acros
223120025
2.5 kg Plastic bottle
£574.00
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1313-99-1
74.69
GNRSAWUEBMWBQH-UHFFFAOYSA-N
14805
O=[Ni]
NiO
MFCD00011145
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
oxonickel

Specifications

1960°C
Black-Gray
NiO
MFCD00011145
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
75 to 77% (Ni)
6.6700g/mL
1313-99-1
Powder
NiO
15, 6597
Solubility in water: insoluble in water. Other solubilities: insoluble in caustic solutions,soluble in acids and ammonium hydroxide
O=[Ni]
74.69
74.7
6.67
Nickel(II) oxide, black, 76% Ni
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer by inhalation.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 215-215-7

missing translation for 'rtecsNumber' : QR8400000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only