missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, 99% (metals basis)

Product Code. p-4407671
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11341739

Brand: Thermo Scientific Alfa Aesar 012359.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

It is used in nickel-iron battery (Edison Battery) and fuel cells. It is a precursor for many nickel salts and nickel alloys including nickel-steel allys, for example invar and elinvar. It is used in electrochromic devices, frits, ferrites, and porcelain glazes. It also serves as a hydrogenation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% (metals basis)
Solubility Information Insoluble in water; soluble in acids and ammonium hydroxide.
Formula Weight 74.71
Density 6.67 g/mL
Chemical Name or Material Nickel(II) oxide
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) oxide
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Carcinogenicity Category 1A
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H372-Causes damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Nickel oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.