missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) oxide, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
It is used in nickel-iron battery (Edison Battery) and fuel cells. It is a precursor for many nickel salts and nickel alloys including nickel-steel allys, for example invar and elinvar. It is used in electrochromic devices, frits, ferrites, and porcelain glazes. It also serves as a hydrogenation catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1984°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Insoluble in water; soluble in acids and ammonium hydroxide. |
Formula Weight | 74.71 |
Density | 6.67 g/mL |
Chemical Name or Material | Nickel(II) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'