missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, Puratronic™, 99.998% (metals basis)

Product Code. 11326947
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326947 25 g 25g
11316947 5 g 5g
11336947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326947 Supplier Thermo Scientific Alfa Aesar Supplier No. 010819.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

Nickel (II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in ammonium hydroxide. Insoluble in water.
Formula Weight 74.71
Density 6.67
Chemical Name or Material Nickel(II) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.