missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)

Product Code. 11375057
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375057 25 g 25g
11365057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375057 Supplier Thermo Scientific Alfa Aesar Supplier No. 040757.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126956-48-7 | C2NiO4 | 146.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126956-48-7
Molecular Formula C2NiO4
Molecular Weight (g/mol) 146.71
MDL Number MFCD00167155
InChI Key DOLZKNFSRCEOFV-UHFFFAOYSA-L
Synonym nickel ii oxalate hydrate, acmc-1c3o8, nickel 2+ hydrate oxalate, nickel ii oxalate hydrate, puratronic, nickel 2+ ion hydrate oxalate, nickel 2+ ethanedioate-water 1/1/1, nickel, ethanedioato 2--ko1,ko2-, hydrate 9ci
PubChem CID 16213068
SMILES [Ni++].[O-]C(=O)C([O-])=O

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NiC2O4·xH2O (x≈2)
UN Number UN3077
Merck Index 14,6514
IUPAC Name nickel(2+) oxalate
Formula Weight 146.73 (Anhydrous)
Percent Purity 99.9985%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Nickel(II) oxalate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.