missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxalate dihydrate

Product Code. 11398248
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398248 50 g 50g
11308258 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398248 Supplier Thermo Scientific Alfa Aesar Supplier No. 039454.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6018-94-6 | C2H4NiO6 | 182.741 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-94-6
Molecular Formula C2H4NiO6
Molecular Weight (g/mol) 182.741
MDL Number MFCD00167155
InChI Key VNYOIRCILMCTHO-UHFFFAOYSA-L
Synonym nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
PubChem CID 516789
IUPAC Name nickel(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]

Specifications

Odor Odorless
Quantity 50 g
UN Number UN3077
Merck Index 14,6514
Solubility Information Soluble in acids; Insoluble in water
Formula Weight 182.76 (146.73 Anhydrous)
Physical Form Powder
Chemical Name or Material Nickel(II) oxalate dihydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) oxalate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Nickel(II) oxalate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.