missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxalate dihydrate

Product Code. 11308258
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11308258

Brand: Thermo Scientific Alfa Aesar 039454.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6018-94-6 | C2H4NiO6 | 182.741 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-94-6
Molecular Formula C2H4NiO6
Molecular Weight (g/mol) 182.741
MDL Number MFCD00167155
InChI Key VNYOIRCILMCTHO-UHFFFAOYSA-L
Synonym nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
PubChem CID 516789
IUPAC Name nickel(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]

Specifications

Odor Odorless
Quantity 250 g
UN Number UN3077
Merck Index 14,6514
Solubility Information Soluble in acids; Insoluble in water
Formula Weight 182.76 (146.73 Anhydrous)
Physical Form Powder
Chemical Name or Material Nickel(II) oxalate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.