missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxalate dihydrate, Thermo Scientific Chemicals

Catalog Number p-7050785
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
6018-94-6
C2H4NiO6
182.741
MFCD00167155
VNYOIRCILMCTHO-UHFFFAOYSA-L
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
516789
nickel(2+);oxalate;dihydrate
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]
This item is not returnable. View return policy
6018-94-6
C2H4NiO6
182.741
MFCD00167155
VNYOIRCILMCTHO-UHFFFAOYSA-L
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
516789
nickel(2+);oxalate;dihydrate
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 50 g
UN Number UN3077
Merck Index 14,6514
Solubility Information Soluble in acids; Insoluble in water
Formula Weight 182.76 (146.73 Anhydrous)
Physical Form Powder
Chemical Name or Material Nickel(II) oxalate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'