missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) octanoate, in mineral spirits (8% Ni)

Product Code. 11368278
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11368278

Brand: Thermo Scientific Alfa Aesar 039479.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4995-91-9 | C8H16NiO2 | 202.907 g/mol

Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4995-91-9
Molecular Formula C8H16NiO2
Molecular Weight (g/mol) 202.907
MDL Number MFCD00064239
InChI Key ANYMYNDNCKURBB-UHFFFAOYSA-N
Synonym nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
PubChem CID 54611835
IUPAC Name nickel;octanoic acid
SMILES CCCCCCCC(=O)O.[Ni]

Specifications

Color Green
Density 0.95 g/mL
Flash Point 40°C (104°F)
Linear Formula [CH3(CH2)6COO]2Ni
Quantity 25 g
UN Number UN1993
Solubility Information Miscible with ethanol,chloroform,acetonitrile,ether,carbon disulfide,petroleum ether and glacial acetic acid. Immiscible with water.
Formula Weight 345.1
Concentration or Composition (by Analyte or Components) 8% Ni
Physical Form Liquid
Chemical Name or Material Nickel(II) octanoate, In mineral spirits
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.