missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min

Product Code. 11326638
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326638 500 g 500g
11336638 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326638 Supplier Thermo Scientific Alfa Aesar Supplier No. 012222.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel plating, manufacturing of ceramic colors | CAS: 13478-00-7 | H12N2NiO12 | 290.79 g/mol

Nickel plating, manufacturing of ceramic colorsNickel(II) nitrate hexahydrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
IUPAC Name nickel(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 137°C
Melting Point 56.7°C
Color Green
Physical Form Flakes
Quantity 500 g
Linear Formula Ni(NO3)2·6H2O
UN Number UN2725
Merck Index 14,6513
Solubility Information Soluble in water,alcoholVery soluble in water; Soluble in ethanol.
Formula Weight 290.80 (182.70 Anhydrous)
Grade Technical
Concentration or Composition (by Analyte or Components) Ni 19.8% min.
Odor Odorless
Sensitivity Hygroscopic
Density 2.05 g/mL
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302+H332-Harmful if swallowed or if inhaled
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Nickel(II) nitrate,hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.