missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) nitrate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number 10401651
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
13478-00-7, 13138-45-9
H12N2NiO12
290.79
MFCD00149805
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
61630
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
13478-00-7, 13138-45-9
H12N2NiO12
290.79
MFCD00149805
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
61630
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Boiling Point 137.0°C
Melting Point 56.7°C
Color Blue to Green
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ni(NO3)2·6H2O
Merck Index 15,6598
Solubility Information Solubility in water: 940g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
IUPAC Name nickel(2+);dinitrate;hexahydrate
Formula Weight 290.8
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Density 2.05
Chemical Name or Material Nickel(II) nitrate hexahydrate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'