missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) iodide, anhydrous, 99.5% (metals basis)

Product Code. 11348117
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348117 25 g 25g
11358117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348117 Supplier Thermo Scientific Alfa Aesar Supplier No. 022893.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13462-90-3 | I2Ni | 312.502 g/mol

Nickel(II) iodide serves as a catalyst in carbonylation reactions. It is also used as a reagent in organic synthesis, especially in conjunction with samarium(II) iodide. It plays an important role in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-90-3
Molecular Formula I2Ni
Molecular Weight (g/mol) 312.502
MDL Number MFCD00016264
InChI Key BFSQJYRFLQUZKX-UHFFFAOYSA-L
Synonym nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
PubChem CID 26038
IUPAC Name diiodonickel
SMILES [Ni](I)I

Specifications

Melting Point 797°C
Quantity 25 g
UN Number UN3077
Solubility Information Soluble in water and alcohols.
Formula Weight 312.51
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.834 g/mL
Assay (metals basis)
Chemical Name or Material Nickel(II) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.