missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11358117
Click to view available options
Quantity:
5 g
25 g

Nickel(II) iodide serves as a catalyst in carbonylation reactions. It is also used as a reagent in organic synthesis, especially in conjunction with samarium(II) iodide. It plays an important role in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-90-3
Molecular Formula I2Ni
Molecular Weight (g/mol) 312.502
MDL Number MFCD00016264
InChI Key BFSQJYRFLQUZKX-UHFFFAOYSA-L
Synonym nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
PubChem CID 26038
IUPAC Name diiodonickel
SMILES [Ni](I)I

Specifications

Melting Point 797°C
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in water and alcohols.
Formula Weight 312.51
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.834 g/mL
Assay (metals basis)
Chemical Name or Material Nickel(II) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'