missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) hydroxide, Ni 61%

Product Code. 11391839
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391839 5 g 5g
11381839 250 g 250g
11311849 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391839 Supplier Thermo Scientific Alfa Aesar Supplier No. 012517.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12054-48-7 | H2NiO2 | 92.71 g/mol

Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-48-7
Molecular Formula H2NiO2
Molecular Weight (g/mol) 92.71
MDL Number MFCD00011140
InChI Key BFDHFSHZJLFAMC-UHFFFAOYSA-L
IUPAC Name nickel(2+) dihydroxide
SMILES [OH-].[OH-].[Ni++]

Specifications

Melting Point 230°C (decomposition)
Color Green
Physical Form Powder
Quantity 5 g
Linear Formula Ni(OH)2
UN Number UN3077
Formula Weight 92.7
Concentration or Composition (by Analyte or Components) Ni 61%
Odor Odorless
Density 4.1g/mL
Chemical Name or Material Nickel(II) hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.