missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) formate dihydrate

Product Code. 11331349
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331349 2 kg 2kg
11321349 100 g 100g
11311349 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331349 Supplier Thermo Scientific Alfa Aesar Supplier No. 013801.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Nickel Catalyst Preparations | CAS: 15694-70-9 | Ni(HCO2)2·2H2O

Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15694-70-9
Molecular Formula Ni(HCO2)2·2H2O
MDL Number MFCD00153034

Specifications

Melting Point 130°C to 140°C-2H2O
Color Green
Physical Form Powder
Quantity 2 kg
Solubility Information In the range 180-200° decomposes to Ni,CO,CO2,H2,H2O,CH4. Partially soluble in water
Formula Weight 184.77 (148.74anhydrous)
Appearance Green powder
Density 2.15
Chemical Name or Material Nickel(II) formate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.