missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) fluoride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog Number 11392158
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
10028-18-9
MFCD00016263
This item is not returnable. View return policy
10028-18-9
MFCD00016263

Nickel fluoride comprises the passivating surface that forms on nickel alloys, e.g. monel, which is why such materials are good to store or transport hydrogen fluoride or elemental fluorine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Crystalline
Quantity 10 g
UN Number UN3288
Merck Index 14,6508
Solubility Information Slightly soluble in water
Formula Weight 96.71
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 4.72 g/mL
Chemical Name or Material Nickel(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'