missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) cyanide tetrahydrate

Product Code. 11388278
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388278 50 g 50g
11378278 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388278 Supplier Thermo Scientific Alfa Aesar Supplier No. 039480.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Metallurgy, Nickel-Plating | CAS: 13477-95-7 | C2H8N2NiO4 | 182.789 g/mol

Applied in metallurgy, nickel-plating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-95-7
Molecular Formula C2H8N2NiO4
Molecular Weight (g/mol) 182.789
MDL Number MFCD00049490
InChI Key VHHVAACTKRUWEK-UHFFFAOYSA-N
Synonym nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate
PubChem CID 71317429
IUPAC Name nickel(2+);dicyanide;tetrahydrate
SMILES [C-]#N.[C-]#N.O.O.O.O.[Ni+2]

Specifications

Melting Point 200°C-4H2O
Color Green
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water. Soluble in ammonium carbonate,ammonia,alkali cyanides
Formula Weight 182.73 (110.73 Anhydrous)
Appearance Light green powder
Chemical Name or Material Nickel(II) cyanide tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.