missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) citrate hydrate, 98%

Product Code. p-7050886
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11354587 100 g 100g
11364587 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11354587

Brand: Thermo Scientific Alfa Aesar 039634.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6018-92-4 | C12H10Ni3O14 | 554.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-92-4
Molecular Formula C12H10Ni3O14
Molecular Weight (g/mol) 554.28
MDL Number MFCD00054366
InChI Key UPPLJLAHMKABPR-UHFFFAOYSA-H
Synonym nickel ii citrate hydrate
PubChem CID 131881812
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;nickel
SMILES [Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 100 g
UN Number UN3077
Formula Weight 554.33 (Anhydrous)
Percent Purity ≥98%
Physical Form Chunks and/or Powder
Chemical Name or Material Nickel(II) citrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.