missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11346937
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346937 25 g 25g
11356937 5 g 5g
11336937 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346937 Supplier Thermo Scientific Alfa Aesar Supplier No. 010815.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69098-15-3 | Cl2Ni | 129.59 g/mol

It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69098-15-3
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.59
MDL Number MFCD00149808
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
Synonym nickel ii chloride hydrate, nickel chloride hydrate, cl2ni.h2o, nicl2.h2o, nickel 11 chloride hydrate, nickel ii chloride hydrate, puratronic, nickel ii chloride hydrate trace metals basis
PubChem CID 16212333
SMILES [Cl-].[Cl-].[Ni++]

Specifications

Color Green
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula NiCl2·xH2O
UN Number UN3288
Merck Index 14,6505
Solubility Information Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide.
IUPAC Name dichloronickel;hydrate
Formula Weight 129.60 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Nickel(II) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.