missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)

Product Code. 10137753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137753 100 g Glass bottle 100g
10626592 5 g Glass bottle 5g
10696772 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137753 Supplier Thermo Scientific Acros Supplier No. 193571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0 , 7718-54-9
Molecular Formula Cl2Ni·6H2O
Molecular Weight (g/mol) 237.71
MDL Number MFCD00149809

Specifications

CAS Min % 100.0
CAS Max % 100.0
Physical Form Solution
Quantity 100 g
Assay Percent Range 99.999%
Linear Formula NiCl2·6H2O
Fieser 14,351; 15,96; 17,86
Merck Index 15, 6590
Solubility Information Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Formula Weight 237.71
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Nickel(II) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.