Learn More
Nickel(II) chloride, anhydrous, 99% (metals basis)
Description
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1001°C |
| Color | Yellow to Orange |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 99% (metals basis) |
| Solubility Information | Soluble in water,ethylene glycol,ethanol and ammonium hydroxide. Insoluble in ammonia and nitric acid. |
| Formula Weight | 129.6 |
| Sensitivity | Hygroscopic |
| Appearance | Yellow to orange powder |
| Vapor Pressure | 1.33 hPa (1mm Hg) at 20°C |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.