missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, anhydrous, 99.99% (metals basis)

Product Code. 11325747
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325747 25 g 25g
11315747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325747 Supplier Thermo Scientific Alfa Aesar Supplier No. 043441.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol

Nickel(II) chloride is used in electroplating, electroforming ,electroless plating, tea and coffee plantation, dye mordant. It is also used in semiconducting metal-containing polymer synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Tekniske data

Melting Point 1,001°C
Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NiCl2
UN Number UN3288
Merck Index 14,6505
Solubility Information It is soluble in water,ethylene glycol,ethanol, ammonium hydroxide. It is insoluble in ammonia, nitric acid.
Formula Weight 129.6
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.55 g/mL
Assay (metals basis)
Chemical Name or Material Nickel (II) chloride, Anhydrous
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.