missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis)

Product Code. 11359237
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359237 1 kg 1kg
11369237 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359237 Supplier Thermo Scientific Alfa Aesar Supplier No. 040106.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel Plating pH Adjuster, Catalysts, Ceramic Colors and Glazes | CAS: 155775-31-8 | CH4Ni3O7 | 304.12 g/mol

Nickel plating pH adjuster, catalysts, ceramic colors and glazes.Nickel(II) carbonate (basic) hydrate is used as a precursor to catalysts and in ceramic applications. It acts as an intermediate in the hydrometallurgical purification of nickel from its ores. It is also used in electroplating of nickel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155775-31-8
Molecular Formula CH4Ni3O7
Molecular Weight (g/mol) 304.12
MDL Number MFCD00016262
InChI Key ATYNVYNPWGEMMI-UHFFFAOYSA-H
Synonym nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis
PubChem CID 86176052
IUPAC Name trinickel(2+) tetrahydroxide carbonate
SMILES [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Color Green
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 304.18 (Anhydrous)
Odor Odorless
Appearance Green powder
Chemical Name or Material Nickel(II) carbonate (basic) hydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) carbonate (basic) hydrate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P201-Obtain special instructions before use.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.