missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide hydrate, 98%, for analysis

Product Code. 10082700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082700 100 g Glass bottle 100g
10398200 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082700 Supplier Thermo Scientific Acros Supplier No. 214871000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207569-11-7 | Br2Ni | 218.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207569-11-7 , 13462-88-9
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.50
MDL Number MFCD00149806
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci
PubChem CID 57377118
SMILES [Ni++].[Br-].[Br-]

Specifications

Melting Point 963.0°C
Color Orange
CAS Min % 97.5
CAS Max % 100.0
pH 5 to 7 (5% aq. soln.)
Physical Form Crystalline Powder or Needles
Quantity 100 g
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Linear Formula NiBr2·xH2O
Fieser 04,351
Merck Index 10, 6340
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether
IUPAC Name dibromonickel;hydrate
Formula Weight 218.53
Percent Purity 98%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Nickel(II) bromide hydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) bromide hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H341-Suspected of causing genetic defects.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.