missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide, 99%

Product Code. 10096652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096652 10 g Glass bottle 10g
10276590 50 g Glass bottle 50g
10698824 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096652 Supplier Thermo Scientific Acros Supplier No. 203360100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13462-88-9 | Br2Ni | 218.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-88-9
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.53
MDL Number MFCD00011141
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
PubChem CID 278492
IUPAC Name dibromonickel
SMILES [Ni](Br)Br

Specifications

Melting Point 963.0°C
Color Brown
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder and/or Chunks
Quantity 10 g
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Linear Formula NiBr2
Fieser 04,351
Merck Index 15, 6587
Solubility Information Solubility in water: 567g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether
Formula Weight 218.53
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Nickel(II) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) bromide
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H341-Suspected of causing genetic defects.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.