missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)

Product Code. 11400131
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400131 100 x 100 mm Each
11490121 50 x 50 mm Each
11410131 100 x 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400131 Supplier Thermo Scientific Alfa Aesar Supplier No. 044822.GH
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.5.6g/50 x 50mm | CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray foil
Quantity 100 x 100 mm
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
Form Foil
Chemical Name or Material Nickel
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Nickel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.