Learn More
Nickel Copper Iron Manganese rod, 1.27cm (0.5 in.) dia., Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 036457.BS
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel Copper Iron Manganese rod | |
Ni:Cu:Fe:Mn; 63:28 to 34:2.5 max:2 max wt% | |
10 cm | |
Rod |
1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164 | |
MFCD02091727 | |
≈104g/10cm |
Safety and Handling
- Nickel Copper Iron Manganese rod
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
- Carcinogenicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .