Learn More
Nickel Copper foil, alloy 400, 0.51mm (0.02in) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045148.HB
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Nickel Copper foil, alloy 400 finds application in marine engineering, chemical and hydro-carbon processings, heat exchangers, valves and pumps. Large use of it is made in the alkylation units, namely in the reacting section in contact with concentrated hydrofluoric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Copper | |
Foil | |
MFCD00801098 | |
Insoluble in water. |
Alloy 400 | |
Ni:Cu; 67:33 wt.% | |
300 x 300 mm | |
Foil |
RUO – Research Use Only