missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium, sputtering target, 76.2in (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis), Thermo Scientific™

Product Code. 15494805
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494805 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494805 Supplier Thermo Scientific Alfa Aesar Supplier No. 040872.KS
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 11106-97-1
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 1 Ea.
Odor Odorless
Molecular Formula CrNi
MDL Number MFCD00673110
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES [Cr].[Ni]
IUPAC Name chromium;nickel
Molecular Weight (g/mol) 110.69
PubChem CID 11320920
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Nickel Chromium, sputtering target, 76.2in (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.