missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium lump, 99.9% (metals basis)

Product Code. 11343378
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343378 100 g 100g
11353378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343378 Supplier Thermo Scientific Alfa Aesar Supplier No. 036298.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition | CAS: 11106-97-1 | CrNi | 110.69 g/mol

For vacuum deposition. hey are also used in some dental restorations. Used as Heating Elements, Thermocouples, Wear Resistant Alloys. Used as intermediate and research. Nichrome wire is commonly used in ceramics and Pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
IUPAC Name chromium nickel
SMILES [Cr].[Ni]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 100 g
Odor Odorless
Particle Size 3 to 12 mm
Form Sintered Lump
Solubility Information Insoluble in water.
Percent Purity 99.9%
Assay Metals basis
Chemical Name or Material Nickel Chromium lump

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.