Learn More
Nickel Chromium Iron powder, -325 mesh, Thermo Scientific Chemicals
Description
Nickel Chromium Iron powder is used for process piping, heat exchangers, carburizing equipment, heating-element sheathing, and nuclear steam-generator tubing, slitting, leveling & tension leveling, shearing & slearing, plate burning, annealing, turning & polishing, warehousing & repackaging, painting, chemical, automotive, aircraft & aerospace, medical, mechanical & food & beverage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nickel Chromium Iron powder |
| Quantity | 1 kg |
| Molecular Formula | Ni:Cr:Fe; 58:27-31:7-11 wt% |
| MDL Number | MFCD01741927 |
| UN Number | UN3089 |
| Solubility Information | Insoluble in water |
| Packaging | Glass bottle |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.