missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride Hexahydrate, TRC

Product Code. 30481669
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481669 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481669 Supplier Toronto Research Chemicals Supplier No. TRCN3951005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Chloride Hexahydrate
CAS 7791-20-0
Molecular Formula Cl2Ni 6(H2O)
Quantity 5 g
Synonym Nickel Chloride (NiCl2) Hydrate, Nickel Chloride Hexahydrate, Nickel Chloride Hexahydrate, Nickel Dichloride Hexahydrate, Nickel(2+) Chloride Hexahydrate, Nickel(II) Chloride Hexahydrate
InChI Formula InChI=1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ni+2]
IUPAC Name nickel(2+);dichloride;hexahydrate
Molecular Weight (g/mol) 237.69
Formula Weight 235.9364
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.