missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Specifications
Specifications
| CAS | 7791-20-0 |
| Physical Form | Crystalline |
| Quantity | 1 kg |
| Molecular Formula | NiCl2 · 6H2O |
| Formula Weight | 237.7 |
| Chemical Name or Material | Nickel Chloride Hexahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction