missing translation for 'onlineSavingsMsg'
Learn More

Nickel ammonium sulfate hexahydrate

Product Code. p-4824827
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11341849

Brand: Thermo Scientific Alfa Aesar 012519.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7785-20-8 | Ni(NH4)2(SO4)2·6H2O

It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-20-8
Molecular Formula Ni(NH4)2(SO4)2·6H2O
MDL Number MFCD00150260
Synonym Ammonium nickel sulfate hexahydrate

Specifications

Color Green
Physical Form Large Crystals
Quantity 100 g
Solubility Information Soluble in ammonium sulphate. Insoluble in alcohol. Slightly soluble in water.
Formula Weight 394.99 (286.90 Anhydrous)
Odor Odorless
Appearance Green crystals
Density 1.923
Chemical Name or Material Nickel ammonium sulfate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.