missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel ammonium sulfate hexahydrate, Thermo Scientific Chemicals
166.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green |
Physical Form | Large Crystals |
Quantity | 500 g |
Solubility Information | Soluble in ammonium sulphate. Insoluble in alcohol. Slightly soluble in water. |
Formula Weight | 394.99 (286.90 Anhydrous) |
Odor | Odorless |
Appearance | Green crystals |
Density | 1.923 |
Chemical Name or Material | Nickel ammonium sulfate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'