missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel aluminum, 99.0% min (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Gray |
Appearance | Gray powder, Intermetallic |
Assay Percent Range | 99.0% min. (metals basis) |
Quantity | 10 g |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Nickel aluminum |
Safety and Handling
Product Identifier
- Nickel aluminum
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'