missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Fe/Co, Alloy 909, N19909, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Fe/Co, Alloy 909, N19909, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Boron/Phosphorus/Nitrogen/Oxygen/Carbon/Tantalum/Copper/Aluminium/Tungsten/Chromium/Manganese/Silicon/Titanium/Niobium/Cobalt/Nickel/Iron |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.00012 %/Boron 0.0013 %/Phosphorus 0.002 %/Nitrogen 0.0026 %/Oxygen 0.004 %/Carbon 0.006 %/Tantalum 0.006 %/Copper 0.007 %/Aluminium 0.009 %/Tungsten 0.009 %/Chromium 0.01 %/Manganese 0.03 %/Silicon 0.42 %/Titanium 1.62 %/Niobium 4.6 %/Cobalt 13.1 %/Nickel 37.7 %/Iron 42.4 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction