missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo/W, Alloy 686, N06686, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo/W, Alloy 686, N06686, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Tantalum/Boron/Calcium/Arsenic/Lanthanum/Tin/Sulfur/Oxygen/Zirconium/Phosphorus/Carbon/Magnesium/Vanadium/Nitrogen/Cobalt/Silicon/Copper/Titanium/Niobium/Manganese/Aluminium/Iron/Tungsten/Molybdenum/Chromium/Nickel/Lead/Neodymium/Silver/Yttrium/Hafnium/Zinc/Antimony/Bismuth/Rhenium/Cadmium |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tantalum -100 ppm/Boron -20 ppm/Calcium -20 ppm/Arsenic -10 ppm/Lanthanum -7 ppm/Tin -6 ppm/Sulfur 0.0005 %/Oxygen 0.0006 %/Zirconium 0.0013 %/Phosphorus 0.003 %/Carbon 0.0087 %/Magnesium 0.009 %/Vanadium 0.009 %/Nitrogen 0.0097 %/Cobalt 0.017 %/Silicon 0.037 %/Copper 0.039 %/Titanium 0.058 %/Niobium 0.13 %/Manganese 0.27 %/Aluminium 0.34 %/Iron 0.44 %/Tungsten 3.59 %/Molybdenum 16.1 %/Chromium 20.4 %/Nickel 58.6 %/Lead (<10) ppm/Neodymium (<10) ppm/Silver (<10) ppm/Yttrium (<10) ppm/Hafnium (<100) ppm/Zinc (<100) ppm/Antimony (<20) ppm/Bismuth (<20) ppm/Rhenium (<3) ppm/Cadmium (<30) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction