missing translation for 'onlineSavingsMsg'
Learn More

Nickel AA Standard: Ni @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17977349
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977349 500 mL 500mL
17969219 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17977349 Supplier VHG Labs Supplier No. VHGAANIN500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto contiene un precursor de explosivos de acuerdo con el Reglamento (CE) No 1148/2019, todos los clientes deben completar una declaración antes de la entrega.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nickel AA Standard: Ni @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Nickel
CAS 7440-02-0
Methods AA/GFAA
Analyte or Component Names Nickel
Shipping Condition Room Temperature
Molecular Formula Ni
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Raney alloy, Fibrex, Nickel catalyst, Nickel particles, Raney-Nickel
InChI Formula InChI=1S/Ni
SMILES [Ni]
IUPAC Name nickel
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 58.69
Quantity 500 mL
Formula Weight 57.9353 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.