missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nicastrin Recombinant Rabbit Monoclonal Antibody (002), FITC

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA540999
This item is not returnable.
View return policy
Description
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
NCSTN (Nicastrin) is a gene that encodes a type I transmembrane glycoprotein that is an integral component of the multimeric gamma-secretase complex. The encoded protein cleaves integral membrane proteins, including Notch receptors and beta-amyloid precursor protein, and may be a stabilizing cofactor required for gamma-secretase complex assembly. The cleavage of beta-amyloid precursor protein yields amyloid beta peptide, the main component of the neuritic plaque and the hallmark lesion in the brains of patients with Alzheimer's disease; however, the nature of the encoded protein's role in Alzheimer's disease is not known for certain. Mutations in this gene are associated with familial acne inversa. A pseudogene of this gene is present on chromosome 21.
Specifications
| Nicastrin | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| Q92542 | |
| NCSTN | |
| Recombinant Human Nicastrin/NCSTN protein. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 002 | |
| FITC | |
| NCSTN | |
| 9430068N19Rik; AA727311; anterior pharynx-defective 2; Aph2; ATAG1874; D1Dau13e; Kiaa0253; mKIAA0253; NCSTN; Nct; Nicastrin; OTTHUMP00000031840; UNQ1874/PRO4317 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23385 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction