missing translation for 'onlineSavingsMsg'
Learn More

Nicardipine hydrochloride, MedChemExpress

Product Code. 16487947
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487947 5 g 5g
16441269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487947 Supplier Medchem Express Supplier No. HY12515A
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nicardipine hydrochloride (YC-93) is a calcium channel blocker with an IC50 of 1 μM for blocking cardiac calcium channels. Nicardipine hydrochloride acts as an agent for chronic stable angina and for controlling blood pressure.

Nicardipine hydrochloride (YC-93) is a calcium channel blocker with an IC50 of 1 μM for blocking cardiac calcium channels. Nicardipine hydrochloride acts as an agent for chronic stable angina and for controlling blood pressure.

Specifications

Chemical Name or Material Nicardipine hydrochloride
CAS 54527-84-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H30ClN3O6
Quantity 5 g
Synonym YC-93
Solubility Information DMSO : ≥ 35 mg/mL (67.83 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(OCCN(C)CC2=CC=CC=C2)=O)C1C3=CC=CC([N+]([O-])=O)=C3)OC.[H]Cl
Molecular Weight (g/mol) 515.99
Formula Weight 515.99
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.